Learn More
Medchemexpress LLC Cy3 NHS ester | 1393363-07-9 | 99.2% | 590.15 | 10 MG
SDP
Catalog No. 5000437825
Encompass_Preferred
Cy3 NHS ester is a CY dye, a compound with two nitrogen atoms linked by an odd number of methyl units. Cyanine compounds are characterized by long wavelengths, adjustable absorption and emission, high extinction coefficients, good water solubility, and relatively simple synthesis. These dyes are frequently used for labeling proteins, antibodies, and small molecular compounds.
  • Suitable for protein antibody labeling through a simple mixing reaction
  • Features long wavelength and adjustable absorption
  • Offers high extinction coefficient and good water solubility
  • Available in various quantities for research needs
Catalog No. 50-004-37825 Supplier Medchemexpress LLC Supplier No. HY11249810MG
Cy3 NHS ester is a CY dye, a compound with two nitrogen atoms linked by an odd number of methyl units. Cyanine compounds are characterized by long wavelengths, adjustable absorption and emission, high extinction coefficients, good water solubility, and relatively simple synthesis. These dyes are frequently used for labeling proteins, antibodies, and small molecular compounds.
  • Suitable for protein antibody labeling through a simple mixing reaction
  • Features long wavelength and adjustable absorption
  • Offers high extinction coefficient and good water solubility
  • Available in various quantities for research needs