Learn More
Medchemexpress LLC Cy3 NHS ester | 1393363-07-9 | 99.24% | 590.15 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112498
Encompass_Preferred
Cy3 NHS ester is a CY dye, which stands for Cyanine. Cyanine compounds are characterized by having two nitrogen atoms connected by an odd number of methyl units. These dyes are known for their long wavelength, adjustable absorption and emission, high extinction coefficient, good water solubility, and relatively simple synthesis. CY dyes are frequently used for labeling proteins, antibodies, and small molecular compounds. Labeling protein antibodies can be achieved through a straightforward mixing reaction.
  • Long wavelength
  • Adjustable absorption and emission
  • High extinction coefficient
  • Good water solubility
  • Relatively simple synthesis
  • Often used for labeling proteins, antibodies, and small molecular compounds
  • Protein antibody labeling can be completed through a simple mixing reaction
Catalog No. 50-004-38821