Learn More
Medchemexpress LLC Cy3 NHS ester | 1393363-07-9 | 99.5% | 590.15 | 50 MG
SDP
Catalog No. 5000437440
Encompass_Preferred
Cy3 NHS ester is a CY dye. CY, short for Cyanine, is a compound consisting of two nitrogen atoms connected by an odd number of methyl units. Cyanine compounds have characteristics of long wavelength, adjustable absorption and emission, high extinction coefficient, good water solubility, and relatively simple synthesis. CY dyes are often used for the labeling of proteins, antibodies, and small molecular compounds. For the labeling of protein antibodies, the combination can be completed through a simple mixing reaction.
  • Long wavelength
  • Adjustable absorption and emission
  • High extinction coefficient
  • Good water solubility
  • Relatively simple synthesis
  • Used for labeling of proteins, antibodies, and small molecular compounds
Catalog No. 50-004-37440 Supplier Medchemexpress LLC Supplier No. HY11249850MG
Cy3 NHS ester is a CY dye. CY, short for Cyanine, is a compound consisting of two nitrogen atoms connected by an odd number of methyl units. Cyanine compounds have characteristics of long wavelength, adjustable absorption and emission, high extinction coefficient, good water solubility, and relatively simple synthesis. CY dyes are often used for the labeling of proteins, antibodies, and small molecular compounds. For the labeling of protein antibodies, the combination can be completed through a simple mixing reaction.
  • Long wavelength
  • Adjustable absorption and emission
  • High extinction coefficient
  • Good water solubility
  • Relatively simple synthesis
  • Used for labeling of proteins, antibodies, and small molecular compounds