Learn More
Apexbio Technology LLC Cy3 NHS ester, 25mg. Cas: MFCD: MFCD28505569. Labeling of amino-groups in biomolecules.
SDP
Catalog No. 501904998
Encompass_Preferred
Sulfo-Cy3 NHS ester is a sulfonated, hydrophilic and highly water soluble dye. For labeling proteins and peptides, no organic co-solvent is needed. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4998 Supplier Apexbio Technology LLC Supplier No. A810725
Sulfo-Cy3 NHS ester is a sulfonated, hydrophilic and highly water soluble dye. For labeling proteins and peptides, no organic co-solvent is needed. Other sizes are available. Please inqury us for quote.