Learn More
Apexbio Technology LLC Cy3 NHS ester, 5mg. Cas: MFCD: MFCD28505569. Labeling of amino-groups in biomolecules.
SDP
Supplier:  Apexbio Technology LLC A81075
Encompass_Preferred
Sulfo-Cy3 NHS ester is a sulfonated, hydrophilic and highly water soluble dye. For labeling proteins and peptides, no organic co-solvent is needed. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4999