Learn More
TARGETMOL CHEMICALS INC Cy3 NHS ester 5MG
SDP
Catalog No. 502878315
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cy3 NHS ester is a fluorescent dye used to label amino groups in proteins, peptides, antibodies, and oligonucleotides. The excitation/emission (Ex/Em) values of Cy3 NHS ester are 550 nm/570 nm. Purity 98.03%
Catalog No. 50-287-8315 Supplier TARGETMOL CHEMICALS INC Supplier No. T189465MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cy3 NHS ester is a fluorescent dye used to label amino groups in proteins, peptides, antibodies, and oligonucleotides. The excitation/emission (Ex/Em) values of Cy3 NHS ester are 550 nm/570 nm. Purity 98.03%