Learn More
Apexbio Technology LLC Cy3 NHS ester(Et)(Synonyms: Cy3 NHS ester, Cy3 NHS Ester (ethyl), Cyanine3 NHS ester, Cyanine 3 NHS ester, Cy3 Mono NHS Ester), 5mg.
SDP
Catalog No. 5000060381
Encompass_Preferred
Cy3 NHS ester(Et) is a small molecule commonly employed in biomedical research as a reactive fluorescent reagent Through its N-hydroxysuccinimide (NHS) ester functionality it readily forms stable covalent linkages with primary amine groups present on proteins peptides and other biomolecules Due to its fluorescent character Cy3 NHS ester(Et) is extensively utilized for labeling biological targets enabling visualization and tracking in fluorescence microscopy and imaging analyses This property facilitates investigations into molecular localization trafficking processes and pathology-associated pathways in various experimental models
Catalog No. 50-000-60381 Supplier Apexbio Technology LLC Supplier No. A87655
Cy3 NHS ester(Et) is a small molecule commonly employed in biomedical research as a reactive fluorescent reagent Through its N-hydroxysuccinimide (NHS) ester functionality it readily forms stable covalent linkages with primary amine groups present on proteins peptides and other biomolecules Due to its fluorescent character Cy3 NHS ester(Et) is extensively utilized for labeling biological targets enabling visualization and tracking in fluorescence microscopy and imaging analyses This property facilitates investigations into molecular localization trafficking processes and pathology-associated pathways in various experimental models