Learn More
Medchemexpress LLC Cy3-peg-biotin | 95.0% | 2000 (average) | 10 MG
SDP
Catalog No. 5000297692
Encompass_Preferred
Cy3-PEG-biotin is a polyethylene glycol conjugate bearing a Cy3 fluorescent dye and a biotin affinity tag; it is designed for fluorescence labeling and capture of biomolecules via biotin-streptavidin interactions for microscopy, flow cytometry, and nucleic acid or protein labeling.
  • Provides Cy3 fluorescence for detection (max emission ≈ 562-570 nm).
  • Biotin tag enables high-affinity capture by streptavidin/avidin.
  • PEG spacer (average MW 2000) improves solubility and reduces steric hindrance.
  • Available in small laboratory quantities suitable for labeling workflows.
Catalog No. 50-002-97692 Supplier Medchemexpress LLC Supplier No. HYD251910MG
Cy3-PEG-biotin is a polyethylene glycol conjugate bearing a Cy3 fluorescent dye and a biotin affinity tag; it is designed for fluorescence labeling and capture of biomolecules via biotin-streptavidin interactions for microscopy, flow cytometry, and nucleic acid or protein labeling.
  • Provides Cy3 fluorescence for detection (max emission ≈ 562-570 nm).
  • Biotin tag enables high-affinity capture by streptavidin/avidin.
  • PEG spacer (average MW 2000) improves solubility and reduces steric hindrance.
  • Available in small laboratory quantities suitable for labeling workflows.