Learn More
Medchemexpress LLC Cy3-PEG2-endo-BCN bromide | >98.0% | 843.93 Da | C47H63BrN4O5 | 10 MG
SDP
Catalog No. 5000287362
Encompass_Preferred
Cy3-PEG2-endo-BCN bromide is a Cyanine 3-derived fluorescent reagent with a PEG2 linker and an endo-BCN reactive moiety, supplied as the bromide salt. It is optimized for catalyst-free strain-promoted alkyne-azide cycloaddition (SPAAC) to label azide-bearing biomolecules for fluorescence imaging, biochemical assays, and probe synthesis.
  • Enables catalyst-free SPAAC click chemistry with azide-functionalized molecules.
  • PEG2 linker improves solubility and reduces steric hindrance in conjugation.
  • Bromide salt form simplifies handling and storage.
  • Strong orange-red fluorescence suitable for imaging and detection.
  • Ideal for preparing fluorescent bioconjugates and probes.
Catalog No. 50-002-87362 Supplier Medchemexpress LLC Supplier No. HY155327B10MG
Cy3-PEG2-endo-BCN bromide is a Cyanine 3-derived fluorescent reagent with a PEG2 linker and an endo-BCN reactive moiety, supplied as the bromide salt. It is optimized for catalyst-free strain-promoted alkyne-azide cycloaddition (SPAAC) to label azide-bearing biomolecules for fluorescence imaging, biochemical assays, and probe synthesis.
  • Enables catalyst-free SPAAC click chemistry with azide-functionalized molecules.
  • PEG2 linker improves solubility and reduces steric hindrance in conjugation.
  • Bromide salt form simplifies handling and storage.
  • Strong orange-red fluorescence suitable for imaging and detection.
  • Ideal for preparing fluorescent bioconjugates and probes.