Learn More
Apexbio Technology LLC Cy3 Rabbit Anti-Goat IgG (H+L) Antibody 1ml
SDP
Catalog No. 5000096950
Encompass_Preferred
Cy3 Rabbit Anti-Goat IgG HplusL Antibody is an affinity-purified polyclonal secondary antibody targeting goat immunoglobulins (IgG both heavy and light chains) It is designed to selectively bind to goat-derived primary antibodies enabling detection and amplification of fluorescence signals Cy3 Rabbit Anti-Goat IgG HplusL Antibody exerts its activity primarily through specific recognition and binding of goat IgG Based on these characteristics Cy3 Rabbit Anti-Goat IgG HplusL Antibody holds research potential in immunofluorescence (IF) immunocytochemistry (ICC) immunohistochemistry (IHC) and flow cytometry (FACS) applications
Catalog No. 50-000-96950 Supplier Apexbio Technology LLC Supplier No. K12151
Cy3 Rabbit Anti-Goat IgG HplusL Antibody is an affinity-purified polyclonal secondary antibody targeting goat immunoglobulins (IgG both heavy and light chains) It is designed to selectively bind to goat-derived primary antibodies enabling detection and amplification of fluorescence signals Cy3 Rabbit Anti-Goat IgG HplusL Antibody exerts its activity primarily through specific recognition and binding of goat IgG Based on these characteristics Cy3 Rabbit Anti-Goat IgG HplusL Antibody holds research potential in immunofluorescence (IF) immunocytochemistry (ICC) immunohistochemistry (IHC) and flow cytometry (FACS) applications