Learn More
Apexbio Technology LLC Cy3 TSA Fluorescence System Kit, 200-600 slides
SDP
Supplier:  Apexbio Technology LLC K1051200
Encompass_Preferred
Tyramine Signal Amplification (TSA) from APExBIO technology increases sensitivity by a factor of 100 and enables detection of low-abundance targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC) and in situ hybridization (FISH) applications. The TSA Fluorescence Kit uses horseradish peroxidase (HRP) to directly catalyze the covalent deposition of the fluorophore adjacent to the immobilized enzyme. The labeling process is so fast (less than ten minutes) and the deposition label can be viewed directly under standard or confocal microscopy. TSA fluorescein can also be used in combination with anti-fluorescein. TSA technology is used for bright field microscopy of enzyme conjugates and suitable chromogenic substrates. The use of TSA reagents results in a significant increase in sensitivity compared to standard assays while maintaining stable specificity and resolution. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8284