Learn More
Apexbio Technology LLC Cy3-UTP,25ul (10mM)
SDP
Catalog No. 502239965
Encompass_Preferred
Cy3-UTP (Cyanine 3-UTP) can replace UTP as a substrate for T7 RNA polymerase and generate labeled probes through in vitro transcription. The resulting assembled mRNA will emit orange fluorescence with the involvement of Cy3-UTPs. In addition, Cy3-labeled mRNA is readily visible under ultraviolet light without further staining after gel electrophoresis[1].Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP .Reference:[1]Kim, H., Park, Y., & Lee, J. B. (2015). Self-assembled Messenger RNA Nanoparticles (mRNA-NPs) for Efficient Gene Expression. Scientific Reports, 5(1). doi:10.1038/srep12737. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-223-9965 Supplier Apexbio Technology LLC Supplier No. B83300.025
Cy3-UTP (Cyanine 3-UTP) can replace UTP as a substrate for T7 RNA polymerase and generate labeled probes through in vitro transcription. The resulting assembled mRNA will emit orange fluorescence with the involvement of Cy3-UTPs. In addition, Cy3-labeled mRNA is readily visible under ultraviolet light without further staining after gel electrophoresis[1].Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP .Reference:[1]Kim, H., Park, Y., & Lee, J. B. (2015). Self-assembled Messenger RNA Nanoparticles (mRNA-NPs) for Efficient Gene Expression. Scientific Reports, 5(1). doi:10.1038/srep12737. Other sizes are also available. Please inqury us for quote.