Learn More
Medchemexpress LLC Cy3-yne (sulfo-cyanine3-alkyne) | 1010386-62-5 | 98.1% | 50 MG
SDP
Catalog No. 5000409767
Encompass_Preferred
CY3-YNE is a dye used for labeling soluble proteins, peptides, and oligonucleotides/DNA. It functions as a click chemistry reagent, containing an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing azide groups.
  • Fluorescent dye
  • Click chemistry reagent
  • Contains alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Suitable for labeling soluble proteins, peptides, and oligonucleotides/DNA
Catalog No. 50-004-09767 Supplier Medchemexpress LLC Supplier No. HYD081850MG
CY3-YNE is a dye used for labeling soluble proteins, peptides, and oligonucleotides/DNA. It functions as a click chemistry reagent, containing an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing azide groups.
  • Fluorescent dye
  • Click chemistry reagent
  • Contains alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Suitable for labeling soluble proteins, peptides, and oligonucleotides/DNA