Learn More
TARGETMOL CHEMICALS INC CY5.5 5MG
SDP
Catalog No. 502578911
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cy5.5 (Sulfo-Cyanine5.5) is a near-infrared fluorescent dye for labeling biomolecules such as peptides proteins and oligonucleotides (Ex=673 nm Em=707 nm)purity: 96%
Catalog No. 50-257-8911 Supplier TARGETMOL CHEMICALS INC Supplier No. TD00915MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cy5.5 (Sulfo-Cyanine5.5) is a near-infrared fluorescent dye for labeling biomolecules such as peptides proteins and oligonucleotides (Ex=673 nm Em=707 nm)purity: 96%