Learn More
Apexbio Technology LLC Cy5.5 azide (non-sulfonated), 5mg. Cas: MFCD: NA. used for Click chemistry to label alkyne-modified oligonucleotides
SDP
Supplier:  Apexbio Technology LLC A81145
Encompass_Preferred
This Cy5.5 is a labeling reagent that ready for the use in Click Chemistry reaction which available as 10 mM solution in DMSO. As Cy5.5 azide is a far-red emitting dye, it can replace Alexa Fluor 680, and DyLight 680. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5013