Learn More
Medchemexpress LLC Cy5.5 DBCO | 1857352-95-4 | C59H58N4O14S4 | 1 MG
SDP
Catalog No. 5000375007
Encompass_Preferred
Cy5.5 DBCO is a click chemistry reagent containing a cycloalkynes group. It serves as a linker of Cyanine5.5 fluorophore. The DBCO group facilitates copper-free biocompatible click chemistry with fast reaction kinetics and good stability.
  • Click chemistry reagent
  • Contains a cycloalkynes group
  • Linker for Cyanine5.5 fluorophore
  • DBCO group enables copper-free biocompatible click chemistry
  • Fast reaction kinetics
  • Good stability
Catalog No. 50-003-75007 Supplier Medchemexpress LLC Supplier No. HY1517201MG
Cy5.5 DBCO is a click chemistry reagent containing a cycloalkynes group. It serves as a linker of Cyanine5.5 fluorophore. The DBCO group facilitates copper-free biocompatible click chemistry with fast reaction kinetics and good stability.
  • Click chemistry reagent
  • Contains a cycloalkynes group
  • Linker for Cyanine5.5 fluorophore
  • DBCO group enables copper-free biocompatible click chemistry
  • Fast reaction kinetics
  • Good stability