Learn More
Apexbio Technology LLC Cy5.5 hydrazide (non-sulfonated), 25mg. Cas: N/A MFCD: NA. carbonyl-reactive dye
SDP
Supplier:  Apexbio Technology LLC A826125
Encompass_Preferred
Cy5.5 hydrazide is a reactive dye for the labeling of biomolecules containing carbonyl groups, such as ketone and aldehydes. This dye pocesses a hydrazide group that can be used to label glycans at their reducing end or other reducing sugars, forming a hydrazone linkage. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5094