Learn More
Lumiprobe CY5.5 NHS ESTER 25MG
Catalog No. NC0164634
Encompass_Preferred
Near infrared cyanine dye which is ideal for bioimaging applications. Amine-reactive dye for the labeling of peptides and amino-oligonucleotides. For the labeling of sensitive proteins, use water-soluble sulfo-Cyanine5 NHS ester.
Catalog No. NC0164634 Supplier Lumiprobe Supplier No. 47020
Near infrared cyanine dye which is ideal for bioimaging applications. Amine-reactive dye for the labeling of peptides and amino-oligonucleotides. For the labeling of sensitive proteins, use water-soluble sulfo-Cyanine5 NHS ester.