Learn More
Jackson Immuno Research Labs CY5-AFFINIPURE DONKEY 0.5 MG
SDP
Catalog No. NC0703423
Encompass_Preferred
Cy5-AffiniPure Donkey Anti-Chicken IgY (IgG) (H+L) (min X Bov,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rb,Rat,Shp Sr Prot)
Catalog No. NC0703423 Supplier Jackson Immuno Research Labs Supplier No. 703175155
Cy5-AffiniPure Donkey Anti-Chicken IgY (IgG) (H+L) (min X Bov,Gt,GP,Sy Hms,Hrs,Hu,Ms,Rb,Rat,Shp Sr Prot)