Learn More
Apexbio Technology LLC Cy5 alkyne (non-sulfonated), 25mg. Cas: MFCD: NA. used for Click chemistry to label any molecule bearing azide group.
SDP
Supplier:  Apexbio Technology LLC A813125
Encompass_Preferred
Cy5 alkyne, with low aqueous solubility is a dye ready for the use in Click Chemistry reaction. Various molecules via Click Chemistry reaction can be attached by this deeply colored, and photostable Cy5 fluorophore. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5042