Learn More
Apexbio Technology LLC Cy5 amine (non-sulfonated), 25mg. Cas: MFCD: NA. can be conjugated with reactive groups such as NHS esters, carboxy groups (after carbodiimide activation), and epoxides.
SDP
Supplier:  Apexbio Technology LLC A814325
Encompass_Preferred
Cy5 amine is a reactive dye which contains amino group. This dye can be coupled with a variety of activated esters and other electrophilic reagents. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5060