Learn More
Apexbio Technology LLC Cy5 azide , 5mg. Cas: MFCD: MFCD31580113. used for Click chemistry to label alkyne-modified oligonucleotides
SDP
Supplier:  Apexbio Technology LLC A81285
Encompass_Preferred
Sulfo-Cy5 azide is a sulfonated, hydrophilic and highly water soluble dye for Click chemistry. Due to this features, the dye can be used for labeling biomolecules, especially useful for the labeling of delicate proteins and proteins prone to denaturation, purely aqueous conditions. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5037