Learn More
Aat Bioquest Cy5 biotin conjugate (5 mg)
SDP
Supplier:  Aat Bioquest 3100
Encompass_Preferred
This bifunctional Cy5 biotin conjugate demonstrates excellent avidin binding and strong fluorescence. This catalog is for 5 mg.
Catalog No. 50-224-9391