Learn More
Apexbio Technology LLC Cy5 Boc-hydrazide (non-sulfonated), 5mg. Cas: MFCD: NA. carbonyl-reactive dye
SDP
Supplier:  Apexbio Technology LLC A81445
Encompass_Preferred
Boc-protected Cy5 hydrazide is a reactive dye for the labeling of aldehydes and ketones with high aqueous solubility. Trifluoroacetic acid or other strong acid can remove the Boc-(tert-butoxycarbonyl) protective group. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5063