Learn More
Apexbio Technology LLC Cy5 carboxylic acid (non-sulfonated),5mg
SDP
Catalog No. 502240080
Encompass_Preferred
Cy5 carboxylic acid is a non-activated carboxylic acid with low aqueous solubility. For coupling with amines, or protein labeling, Cy5 NHS ester and water-soluble sulfo-Cy5 NHS ester is preferred. This labeling dye has limited water solubility. For biomolecule labeling, the labeling reagent has low aqueous solubility, using of organic co-solvent to dissolve this molecular is necessary for efficient reaction. First, Cyanine dye should be dissolved in organic solvent and then added to a solution of biomolecule in appropriate aqueous buffer. In the cells, The Cy5 carboxylic acid label on the dye can track the efficiency of the protein expression [1].Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0080 Supplier Apexbio Technology LLC Supplier No. A81335
Cy5 carboxylic acid is a non-activated carboxylic acid with low aqueous solubility. For coupling with amines, or protein labeling, Cy5 NHS ester and water-soluble sulfo-Cy5 NHS ester is preferred. This labeling dye has limited water solubility. For biomolecule labeling, the labeling reagent has low aqueous solubility, using of organic co-solvent to dissolve this molecular is necessary for efficient reaction. First, Cyanine dye should be dissolved in organic solvent and then added to a solution of biomolecule in appropriate aqueous buffer. In the cells, The Cy5 carboxylic acid label on the dye can track the efficiency of the protein expression [1].Other sizes are also available. Please inqury us for quote.