Learn More
Abcam Cy5 Conjugation Kit (Fast) - Lightning-Link, 100UG
Catalog No. NC2073426
Encompass_Preferred
Cy5 conjugation / labeling in < 20 mins with 30 secs hands-on time using Cy5 Conjugation Kit (Fast) - Lightning-Link ab188288. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2073426 Supplier Abcam Supplier No. AB188288100UG
Cy5 conjugation / labeling in < 20 mins with 30 secs hands-on time using Cy5 Conjugation Kit (Fast) - Lightning-Link ab188288. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement