Learn More
Apexbio Technology LLC EZ Cap™ Cy5 EGFP mRNA (5-moUTP), 100ug (1mg/mL)
SDP
Catalog No. 501998316
Encompass_Preferred
EZ Cap™ Cy5 EGFP mRNA (5-moUTP) will express enhanced green fluorescent protein(EGFP) once entering cells originally isolated from the jellyfish(Aequorea victoria). You can observe the green fluorescence at 509 nm. EGFP is usually used as a reporter for gene regulation and function study. It is applicable in assays for mRNA delivery, translation efficiency, cell viability and in vivo imaging etc. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8316 Supplier Apexbio Technology LLC Supplier No. R10110.1
EZ Cap™ Cy5 EGFP mRNA (5-moUTP) will express enhanced green fluorescent protein(EGFP) once entering cells originally isolated from the jellyfish(Aequorea victoria). You can observe the green fluorescence at 509 nm. EGFP is usually used as a reporter for gene regulation and function study. It is applicable in assays for mRNA delivery, translation efficiency, cell viability and in vivo imaging etc. Other sizes are also available. Please inqury us for quote.