Learn More
Abcam Cy5 Conjugation Kit (Fast) - Lightning-Link, 1MG
Catalog No. NC1271539
Encompass_Preferred
Cy5 conjugation / labeling in < 20 mins with 30 secs hands-on time using Cy5 Conjugation Kit (Fast) - Lightning-Link ab188288. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1271539 Supplier Abcam Supplier No. AB1882881MG
Cy5 conjugation / labeling in < 20 mins with 30 secs hands-on time using Cy5 Conjugation Kit (Fast) - Lightning-Link ab188288. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement