Learn More
Apexbio Technology LLC EZ Cap™ Cy5 Firefly Luciferase mRNA (5-moUTP), 100ug (1mg/mL)
SDP
Catalog No. 501998313
Encompass_Preferred
EZ Cap™ Cy5 Firefly Luciferase mRNA (5-moUTP) will express luciferase protein once entering cells which is initially extracted from firefly Photinus pyralis . This enzyme catalyzes ATP-dependent D-luciferin oxidation and lead to yield chemiluminescence at about 560 nm wavelength. Firefly Luciferase is a frequently used bioluminescent reporter for gene regulation and function study. It is applicable in assays for mRNA delivery, translation efficiency, cell viability and in vivo imaging etc. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8313 Supplier Apexbio Technology LLC Supplier No. R10100.1
EZ Cap™ Cy5 Firefly Luciferase mRNA (5-moUTP) will express luciferase protein once entering cells which is initially extracted from firefly Photinus pyralis . This enzyme catalyzes ATP-dependent D-luciferin oxidation and lead to yield chemiluminescence at about 560 nm wavelength. Firefly Luciferase is a frequently used bioluminescent reporter for gene regulation and function study. It is applicable in assays for mRNA delivery, translation efficiency, cell viability and in vivo imaging etc. Other sizes are also available. Please inqury us for quote.