Learn More
Cayman Chemical Cy5 FIrefly LucIfRase mRNA
SDP
Catalog No. 502945624
Encompass_Preferred
An mRNA for the expression of luciferase; encapsulation in LNPs can be used for mRNA delivery and expression of luciferase protein in vitro
Catalog No. 50-294-5624 Supplier Cayman Chemical Supplier No. 41965100 UG
An mRNA for the expression of luciferase; encapsulation in LNPs can be used for mRNA delivery and expression of luciferase protein in vitro