Learn More
Cayman Chemical Cy5 Firefly Luciferase mRNA (Cap-1; mo5U), 1 mg
SDP
Catalog No. 502945580
Encompass_Preferred
An mRNA for the expression of luciferase; encapsulation in LNPs can be used for mRNA delivery and expression of luciferase protein in vitro
Catalog No. 50-294-5580 Supplier Cayman Chemical Supplier No. 419651 MG
An mRNA for the expression of luciferase; encapsulation in LNPs can be used for mRNA delivery and expression of luciferase protein in vitro