Learn More
Apexbio Technology LLC Cy5 hydrazide (non-sulfonated), 5mg. Cas: MFCD: NA. carbonyl-reactive dye
SDP
Supplier:  Apexbio Technology LLC A81455
Encompass_Preferred
Cy5 hydrazide is a reactive dye which reacts smoothly and nearly quantitatively with various carbonyl groups encountered in biomolecules. The labeling regent is used for the labeling of aldehydes and ketones. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5065