Learn More
Medchemexpress LLC Cy5 maleimide | 1437796-65-0 | 98.0% | 641.24 | 25 MG
SDP
Catalog No. 5000348607
Encompass_Preferred
Cy5 maleimide is a CY dye, a type of cyanine compound. Cyanine compounds are characterized by having two nitrogen atoms connected by an odd number of methyl units. These dyes possess long wavelengths, adjustable absorption and emission, high extinction coefficients, good water solubility, and relatively simple synthesis. CY dyes are frequently used for labeling proteins, antibodies, and small molecular compounds. For protein and antibody labeling, the conjugation can be achieved through a simple mixing reaction.
  • Possesses long wavelengths
  • Adjustable absorption and emission
  • High extinction coefficients
  • Good water solubility
  • Relatively simple synthesis
  • Used for labeling proteins, antibodies, and small molecular compounds
  • Conjugation for protein and antibody labeling can be achieved through simple mixing reaction
Catalog No. 50-003-48607 Supplier Medchemexpress LLC Supplier No. 25MGHY138200
Cy5 maleimide is a CY dye, a type of cyanine compound. Cyanine compounds are characterized by having two nitrogen atoms connected by an odd number of methyl units. These dyes possess long wavelengths, adjustable absorption and emission, high extinction coefficients, good water solubility, and relatively simple synthesis. CY dyes are frequently used for labeling proteins, antibodies, and small molecular compounds. For protein and antibody labeling, the conjugation can be achieved through a simple mixing reaction.
  • Possesses long wavelengths
  • Adjustable absorption and emission
  • High extinction coefficients
  • Good water solubility
  • Relatively simple synthesis
  • Used for labeling proteins, antibodies, and small molecular compounds
  • Conjugation for protein and antibody labeling can be achieved through simple mixing reaction