Learn More
Apexbio Technology LLC Cy5 maleimide (non-sulfonated), 25mg. Cas: MFCD: NA. Labeling of cysteine residues, as well as other thiolated molecules
SDP
Catalog No. 501905052
Encompass_Preferred
Cy5 maleimide is a mono-reactive dye for selectively labeling peptides and proteins which couples with thiol groups. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5052 Supplier Apexbio Technology LLC Supplier No. A813925
Cy5 maleimide is a mono-reactive dye for selectively labeling peptides and proteins which couples with thiol groups. Other sizes are available. Please inqury us for quote.