Learn More
TARGETMOL CHEMICALS INC Cy5-N3 1MG
SDP
Catalog No. 502878495
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. CY5-N3 (Sulfo-Cyanine5-azide) is an anthocyanin fluorescent dye with azide groups, utilized for cell imaging, tissue imaging, and in vivo imaging.
Catalog No. 50-287-8495 Supplier TARGETMOL CHEMICALS INC Supplier No. T189421MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. CY5-N3 (Sulfo-Cyanine5-azide) is an anthocyanin fluorescent dye with azide groups, utilized for cell imaging, tissue imaging, and in vivo imaging.