Learn More
Apexbio Technology LLC Cy5 NHS ester (non-sulfonated), 10mM (in 1mL DMSO)
SDP
Catalog No. 501904991
Encompass_Preferred
Cyanine5 NHS ester is a ideal reactive dye for the labeling of peptides, proteins, and oligonucleotides which contain the amino-groups. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4991 Supplier Apexbio Technology LLC Supplier No. A81025.1
Cyanine5 NHS ester is a ideal reactive dye for the labeling of peptides, proteins, and oligonucleotides which contain the amino-groups. Other sizes are available. Please inqury us for quote.