Learn More
Apexbio Technology LLC Cy5 NHS ester, 25mg. Cas: MFCD: MFCD28142487. Labeling of amino-groups in biomolecules.
SDP
Supplier:  Apexbio Technology LLC A810825
Encompass_Preferred
Sulfo-Cy5 NHS ester is a sulfonated, hydrophilic and highly water soluble dye. For the labeling of various amine-containing molecules in aqueous phase, no organic co-solvent is needed. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5000