Learn More
Medchemexpress LLC Cy5-PEG3-azide | 00-00-0 | C40H55ClN6O4 | 10MG
SDP
Catalog No. 5000258584
Encompass_Preferred
Cy5-PEG3-azide is a Cy5-labeled PEG3 azide reagent for click-chemistry labeling and linker applications. It enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) to attach Cy5 fluorescence to alkyne-bearing molecules for bioconjugation, imaging, and PROTAC synthesis.
  • Provides Cy5 fluorescence for detection and imaging.
  • Contains a PEG3 spacer to improve solubility and reduce steric hindrance.
  • Reactive azide functional group for CuAAC and SPAAC conjugations.
  • Supplied as a dry powder for convenient storage and handling.
  • Store protected from light at -20°C; in solvent, store at -80°C for long-term.
Catalog No. 50-002-58584 Supplier Medchemexpress LLC Supplier No. HY14105810MG
Cy5-PEG3-azide is a Cy5-labeled PEG3 azide reagent for click-chemistry labeling and linker applications. It enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) to attach Cy5 fluorescence to alkyne-bearing molecules for bioconjugation, imaging, and PROTAC synthesis.
  • Provides Cy5 fluorescence for detection and imaging.
  • Contains a PEG3 spacer to improve solubility and reduce steric hindrance.
  • Reactive azide functional group for CuAAC and SPAAC conjugations.
  • Supplied as a dry powder for convenient storage and handling.
  • Store protected from light at -20°C; in solvent, store at -80°C for long-term.