Learn More
Medchemexpress LLC Cy5-PEG3-azide | 00-00-0 | MFCD28142475 | 99.5% | 719.36 | C40H55ClN6O4 | 5 MG
SDP
Catalog No. 5000222765
Encompass_Preferred
Cy5-PEG3-azide is a PEG-based linker dye bearing an azide group for bioorthogonal conjugation. It is designed for use in click chemistry and linker-dependent constructs, enabling efficient attachment to alkyne-bearing partners for bioconjugation and PROTAC synthesis.
  • PEG-based linker with azide functionality for click chemistry.
  • Compatible with copper-catalyzed and strain-promoted azide-alkyne cycloaddition.
  • Suitable for PROTAC synthesis and general bioconjugation workflows.
  • High purity for reliable analytical and preparative results.
  • Store at -20°C and protect from light; in solution, store at -80°C for long-term.
Catalog No. 50-002-22765 Supplier Medchemexpress LLC Supplier No. HY1410585MG
Cy5-PEG3-azide is a PEG-based linker dye bearing an azide group for bioorthogonal conjugation. It is designed for use in click chemistry and linker-dependent constructs, enabling efficient attachment to alkyne-bearing partners for bioconjugation and PROTAC synthesis.
  • PEG-based linker with azide functionality for click chemistry.
  • Compatible with copper-catalyzed and strain-promoted azide-alkyne cycloaddition.
  • Suitable for PROTAC synthesis and general bioconjugation workflows.
  • High purity for reliable analytical and preparative results.
  • Store at -20°C and protect from light; in solution, store at -80°C for long-term.