Learn More
Medchemexpress LLC Cy5-PEG3-endo-BCN | 1881221-50-6 | 98.4% | 869.57 | 1 MG
SDP
Catalog No. 5000359459
Encompass_Preferred
Cy5-PEG3-endo-BCN is a dye derivative of Cyanine 5 (Cy5) that incorporates three PEG units. It features the lyophilic bidentate macrocyclic ligand endo-BCN, which allows for the synthesis of additional macrocyclic complexes. In click chemistry applications, endo-BCN can react with azide-containing molecules to form stable triazoles without the need for catalysts.
  • Dye derivative of Cyanine 5.
  • Contains 3 PEG units.
  • Includes the lyophilic bidentate macrocyclic ligand endo-BCN.
  • Suitable for synthesizing macrocyclic complexes.
  • Reacts with azide groups in click chemistry to form stable triazoles, even in the absence of catalysts.
Catalog No. 50-003-59459 Supplier Medchemexpress LLC Supplier No. HY1496181MG
Cy5-PEG3-endo-BCN is a dye derivative of Cyanine 5 (Cy5) that incorporates three PEG units. It features the lyophilic bidentate macrocyclic ligand endo-BCN, which allows for the synthesis of additional macrocyclic complexes. In click chemistry applications, endo-BCN can react with azide-containing molecules to form stable triazoles without the need for catalysts.
  • Dye derivative of Cyanine 5.
  • Contains 3 PEG units.
  • Includes the lyophilic bidentate macrocyclic ligand endo-BCN.
  • Suitable for synthesizing macrocyclic complexes.
  • Reacts with azide groups in click chemistry to form stable triazoles, even in the absence of catalysts.