Learn More
Medchemexpress LLC Cy5-PEG3-endo-BCN | 1881221-50-6 | 98.4% | 869.57 | 5 MG
SDP
Catalog No. 5000361924
Encompass_Preferred
Cy5-PEG3-endo-BCN is a dye derivative of Cyanine 5 (Cy5) that includes 3 PEG units. It contains the lyophilic bidentate macrocyclic ligand endo-BCN, which can be used to synthesize macrocyclic complexes. In click chemistry, endo-BCN can react with azide-containing molecules to form stable triazoles without the need for catalysts. This product is for research use only.
  • Dye derivative of Cyanine 5 (Cy5)
  • Contains 3 PEG units
  • Includes lyophilic bidentate macrocyclic ligand endo-BCN
  • Enables synthesis of macrocyclic complexes
  • Reacts with azide-containing molecules to form stable triazoles in click chemistry
  • Does not require catalysts for triazole formation
Catalog No. 50-003-61924 Supplier Medchemexpress LLC Supplier No. HY1496185MG
Cy5-PEG3-endo-BCN is a dye derivative of Cyanine 5 (Cy5) that includes 3 PEG units. It contains the lyophilic bidentate macrocyclic ligand endo-BCN, which can be used to synthesize macrocyclic complexes. In click chemistry, endo-BCN can react with azide-containing molecules to form stable triazoles without the need for catalysts. This product is for research use only.
  • Dye derivative of Cyanine 5 (Cy5)
  • Contains 3 PEG units
  • Includes lyophilic bidentate macrocyclic ligand endo-BCN
  • Enables synthesis of macrocyclic complexes
  • Reacts with azide-containing molecules to form stable triazoles in click chemistry
  • Does not require catalysts for triazole formation