Learn More
Medchemexpress LLC Cy5-PEG3-endo-BCN | 1881221-50-6 | 98.4% | 869.57 | C51H69ClN4O6 | 10 MG
SDP
Catalog No. 5000286658
Encompass_Preferred
Cy5-PEG3-endo-BCN is a Cyanine 5 dye derivative containing a three-unit polyethylene glycol spacer and an endo-BCN reactive group. Supplied as a purple to purplish-red solid for research use, it is designed for bioorthogonal labeling of azide-containing biomolecules via catalyst-free strain-promoted azide-alkyne cycloaddition.
  • Dye derivative with PEG3 spacer to improve solubility in aqueous media.
  • Endo-BCN functional group enables catalyst-free click chemistry labeling.
  • High purity suitable for sensitive fluorescence labeling applications.
  • Purple solid appearance for easy visual confirmation.
  • Recommended storage under inert atmosphere at low temperatures.
  • Available in small milligram pack sizes for laboratory research.
Catalog No. 50-002-86658 Supplier Medchemexpress LLC Supplier No. HY14961810MG
Cy5-PEG3-endo-BCN is a Cyanine 5 dye derivative containing a three-unit polyethylene glycol spacer and an endo-BCN reactive group. Supplied as a purple to purplish-red solid for research use, it is designed for bioorthogonal labeling of azide-containing biomolecules via catalyst-free strain-promoted azide-alkyne cycloaddition.
  • Dye derivative with PEG3 spacer to improve solubility in aqueous media.
  • Endo-BCN functional group enables catalyst-free click chemistry labeling.
  • High purity suitable for sensitive fluorescence labeling applications.
  • Purple solid appearance for easy visual confirmation.
  • Recommended storage under inert atmosphere at low temperatures.
  • Available in small milligram pack sizes for laboratory research.