Learn More
Medchemexpress LLC Cy5-PEG5-azide bromide | 98.8% | C44H63BrN6O6 | 2 MG
SDP
Catalog No. 5000355046
Encompass_Preferred
Cy5-PEG5-azide is a PEG-based linker utilized in the synthesis of PROTACs, which are compounds designed to selectively degrade target proteins. This reagent facilitates click chemistry reactions, containing an azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, or strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Enables targeted protein degradation.
  • Supports efficient synthesis of degraders.
  • Compatible with multiple click chemistry methods.
  • Offers a versatile conjugation tool for research.
  • High purity for reliable experimental results.
Catalog No. 50-003-55046 Supplier Medchemexpress LLC Supplier No. HY1410592MG
Cy5-PEG5-azide is a PEG-based linker utilized in the synthesis of PROTACs, which are compounds designed to selectively degrade target proteins. This reagent facilitates click chemistry reactions, containing an azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, or strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Enables targeted protein degradation.
  • Supports efficient synthesis of degraders.
  • Compatible with multiple click chemistry methods.
  • Offers a versatile conjugation tool for research.
  • High purity for reliable experimental results.