Learn More
Apexbio Technology LLC Cy5 Rabbit Anti-Goat IgG (H+L) Antibody 2X1ml
SDP
Catalog No. 5000097064
Encompass_Preferred
Cy5 Rabbit Anti-Goat IgG HplusL Antibody is a fluorescence-labeled secondary antibody targeting goat immunoglobulin G (IgG) specifically binding both heavy and light chains It is designed for signal amplification in immunological assays by binding to primary goat IgG antibodies enabling sensitive detection of target antigens Cy5 Rabbit Anti-Goat IgG HplusL Antibody exerts its biological activity by binding to goat IgG primary antibodies enabling visualization through the Cy5 fluorophore (excitation/emission 650/670 nm) Based on these properties Cy5 Rabbit Anti-Goat IgG HplusL Antibody holds research potential in immunofluorescence staining flow cytometry western blotting and ELISA applications
Catalog No. 50-000-97064 Supplier Apexbio Technology LLC Supplier No. K12162
Cy5 Rabbit Anti-Goat IgG HplusL Antibody is a fluorescence-labeled secondary antibody targeting goat immunoglobulin G (IgG) specifically binding both heavy and light chains It is designed for signal amplification in immunological assays by binding to primary goat IgG antibodies enabling sensitive detection of target antigens Cy5 Rabbit Anti-Goat IgG HplusL Antibody exerts its biological activity by binding to goat IgG primary antibodies enabling visualization through the Cy5 fluorophore (excitation/emission 650/670 nm) Based on these properties Cy5 Rabbit Anti-Goat IgG HplusL Antibody holds research potential in immunofluorescence staining flow cytometry western blotting and ELISA applications