Learn More
Medchemexpress LLC CY5-SE triethylamine salt | 1497420-70-8 | 98.0% | 5 MG
SDP
Catalog No. 5000411065
Encompass_Preferred
Cy5-SE (Cy5 NHS Ester) triethylamine salt is a reactive dye used for labeling amino-groups in peptides, proteins, and oligonucleotides. It is suitable for cost-efficient labeling of soluble proteins, peptides, and oligonucleotides, with an excitation wavelength of 649 nm and emission wavelength of 670 nm.
  • Reactive dye for labeling amino-groups
  • Suitable for cost-efficient labeling of soluble proteins
  • Applicable for peptides and oligonucleotides
  • Excitation wavelength: 649 nm
  • Emission wavelength: 670 nm
Catalog No. 50-004-11065 Supplier Medchemexpress LLC Supplier No. HYD0819A5MG
Cy5-SE (Cy5 NHS Ester) triethylamine salt is a reactive dye used for labeling amino-groups in peptides, proteins, and oligonucleotides. It is suitable for cost-efficient labeling of soluble proteins, peptides, and oligonucleotides, with an excitation wavelength of 649 nm and emission wavelength of 670 nm.
  • Reactive dye for labeling amino-groups
  • Suitable for cost-efficient labeling of soluble proteins
  • Applicable for peptides and oligonucleotides
  • Excitation wavelength: 649 nm
  • Emission wavelength: 670 nm