Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cy5-SE (triethylamine salt) | 1497420-70-8 | 98.0% | 855.07 g/mol | C43H58N4O10S2 | 10 MG
SDP

Catalog No. 5000296843
Encompass_Preferred

CY5-SE (triethylamine salt) is a Cy5-derived N-hydroxysuccinimide (NHS) ester reactive dye for covalent labeling of primary amines in peptides, proteins, oligonucleotides, and small molecules. Supplied as the triethylamine salt, it is compatible with organic solvents and provides strong fluorescence with excitation near 649 nm and emission near 670 nm.

  • Reactive NHS ester for efficient amine conjugation.
  • Triethylamine salt form; soluble in organic solvents.
  • Excitation ~649 nm, emission ~670 nm for strong fluorescence.
  • Molecular weight 855.07 g/mol.
  • Purity 98.0%.
  • Suitable for labeling peptides, proteins, oligonucleotides, and small molecules.

Catalog No. 50-002-96843 Supplier Medchemexpress LLC Supplier No. HYD0819A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CY5-SE (triethylamine salt) is a Cy5-derived N-hydroxysuccinimide (NHS) ester reactive dye for covalent labeling of primary amines in peptides, proteins, oligonucleotides, and small molecules. Supplied as the triethylamine salt, it is compatible with organic solvents and provides strong fluorescence with excitation near 649 nm and emission near 670 nm.

  • Reactive NHS ester for efficient amine conjugation.
  • Triethylamine salt form; soluble in organic solvents.
  • Excitation ~649 nm, emission ~670 nm for strong fluorescence.
  • Molecular weight 855.07 g/mol.
  • Purity 98.0%.
  • Suitable for labeling peptides, proteins, oligonucleotides, and small molecules.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.