Learn More
Apexbio Technology LLC Cy5-UTP, 25ul (10mM). Cas: MFCD: NA. used as a substrate for T7 RNA polymerase and generate labeled probes through in vitro transcription
SDP
Catalog No. 501998343
Encompass_Preferred
Cy5-UTP (Cyanine 5-UTP) can replace UTP as a substrate for T7 RNA polymerase and generate labeled probes through in vitro transcription. The resulting assembled mRNA will emit orange fluorescence with the involvement of Cy5-UTPs. In addition, Cy5-labeled mRNA is readily visible under ultraviolet light without further staining after gel electrophoresis. Probes generated by these methods are suitable for various application, such as FISH, multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with Cy5-UTP. Excitation/Emission (nm): 650/665 nm Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8343 Supplier Apexbio Technology LLC Supplier No. B83330.025
Cy5-UTP (Cyanine 5-UTP) can replace UTP as a substrate for T7 RNA polymerase and generate labeled probes through in vitro transcription. The resulting assembled mRNA will emit orange fluorescence with the involvement of Cy5-UTPs. In addition, Cy5-labeled mRNA is readily visible under ultraviolet light without further staining after gel electrophoresis. Probes generated by these methods are suitable for various application, such as FISH, multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with Cy5-UTP. Excitation/Emission (nm): 650/665 nm Other sizes are also available. Please inqury us for quote.