Learn More
Medchemexpress LLC Cy5-UTP sodium | C45H58N5O22P3S2.xNa | 500 UL
SDP
Catalog No. 5000374119
Encompass_Preferred
Cy5-UTP sodium is a fluorescently labeled ribonucleotide triphosphate that functions as a substrate for terminal deoxynucleotide transferase (TdT). It can be used to label RNA probes generated in vitro (Ex/Em: 650/665 nm). This product is applicable in FISH (Fluorescence In Situ Hybridization) and multi-color fluorescence analysis, particularly in dual-color expression arrays when combined with Cy5-UTP.
  • Fluorescently labeled ribonucleotide triphosphate
  • Substrate for terminal deoxynucleotide transferase (TdT)
  • Can be used to label RNA probes generated in vitro (Ex/Em: 650/665 nm)
  • Applicable in FISH and multi-color fluorescence analysis
  • Suitable for dual-color expression arrays combined with Cy5-UTP
Catalog No. 50-003-74119 Supplier Medchemexpress LLC Supplier No. HY150145A500UL
Cy5-UTP sodium is a fluorescently labeled ribonucleotide triphosphate that functions as a substrate for terminal deoxynucleotide transferase (TdT). It can be used to label RNA probes generated in vitro (Ex/Em: 650/665 nm). This product is applicable in FISH (Fluorescence In Situ Hybridization) and multi-color fluorescence analysis, particularly in dual-color expression arrays when combined with Cy5-UTP.
  • Fluorescently labeled ribonucleotide triphosphate
  • Substrate for terminal deoxynucleotide transferase (TdT)
  • Can be used to label RNA probes generated in vitro (Ex/Em: 650/665 nm)
  • Applicable in FISH and multi-color fluorescence analysis
  • Suitable for dual-color expression arrays combined with Cy5-UTP