Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CY5-YNE (Sulfo-Cyanine5-alkyne) | 1345823-20-2 | 95.29% | 25 MG
SDP

Catalog No. 5000412649
Encompass_Preferred

CY5-YNE (Sulfo-Cyanine5-alkyne) is a reactive dye used for labeling amino-groups in peptides, proteins, and oligonucleotides. It functions as a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have Azide groups.

  • Reactive dye for labeling amino-groups in peptides, proteins, and oligonucleotides.
  • Click chemistry reagent with an Alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.
  • Excitation (Ex): 645 nm.
  • Emission (Em): 670 nm.
  • Appearance: Solid, grayish brown to reddish brown.
  • Storage: 4°C, protect from light.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (protect from light).

Catalog No. 50-004-12649 Supplier Medchemexpress LLC Supplier No. 25MGHYD0820
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CY5-YNE (Sulfo-Cyanine5-alkyne) is a reactive dye used for labeling amino-groups in peptides, proteins, and oligonucleotides. It functions as a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have Azide groups.

  • Reactive dye for labeling amino-groups in peptides, proteins, and oligonucleotides.
  • Click chemistry reagent with an Alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.
  • Excitation (Ex): 645 nm.
  • Emission (Em): 670 nm.
  • Appearance: Solid, grayish brown to reddish brown.
  • Storage: 4°C, protect from light.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.