Learn More
Selleck Chemical LLC Cy7 E2864-25MG
SDP
Catalog No. 5000040515
Encompass_Preferred
Cy7 (Sulfo-Cyanine7) is a fluorescence labelling agent used to label proteins antibodies peptides and oligonucleotides
Catalog No. 50-000-40515 Supplier Selleck Chemical LLC Supplier No. E286425MG
Cy7 (Sulfo-Cyanine7) is a fluorescence labelling agent used to label proteins antibodies peptides and oligonucleotides